1,1-Dibromo-3,3,3-trifluoroacetone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one
001230-5G
001230-25G
001230-100G
FLUO001230-5GEA
0
GBP
FLUO001230-5G
FLUO001230-25G
FLUO001230-100G
1,1-Dibromo-3,3,3-trifluoroacetone
1,1-Dibromo-3,3,3-trifluoroacetone
Formula:
Br₂HCC(O)CF₃ MW: 269.84 g/mol Boiling Pt: 112…113 °C (1013 hPa) Density: 2.188 g/cm³ (20 °C) |
MDL Number:
MFCD00041362 CAS Number: 431-67-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...