1,4-Dibromopentane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001096-10G
001096-50G
FLUO001096-10GEA
0
GBP
FLUO001096-10G
FLUO001096-50G
1,4-Dibromopentane
1,4-Dibromopentane
Formula:
BrH₂C(CH₂)₂BrCHCH₃ MW: 229.94 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: –34 °C Density: 1,687 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000153 CAS Number: 626-87-9 EINECS: 210-967-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...