1,4-Dibromobutane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,4-Dibromo butane
004638-500G
004638-5KG
FLUO004638-500GEA
0
GBP
FLUO004638-500G
FLUO004638-5KG
1,4-Dibromobutane
1,4-Dibromobutane
Formula:
BrH₂C(CH₂)₂CH₂Br MW: 215.92 g/mol Boiling Pt: 197 °C (1013 hPa) Melting Pt: –20 °C Density: 1.83 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000261 CAS Number: 110-52-1 EINECS: 203-775-5 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...