1-Bromotetrafluoro-2-iodoethane ≥97% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Stabilisation:
Stabilised with copper
B21771.06
B21771.14
B21771.22
B21771.06EA
47.3
GBP
B21771.06
B21771.14
B21771.22
1-Bromotetrafluoro-2-iodoethane ≥97% stabilised
1-Bromotetrafluoro-2-iodoethane
Stabilized with copper
Purity: 97%
CAS No.: 421-70-5
Molecular Formula: BrCF2CF2I
Formula Weight: 306.82
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4330
Boiling Point: 78°
Beilstein Reference: 1688466
MDL No.: MFCD00042136
Formula:
BrF₂CCF₂I Boiling Pt: 78 °C Density: 1.648 Storage Temperature: Refrigerator |
MDL Number:
MFCD00042136 CAS Number: 421-70-5 EINECS: 207-008-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...