Citric acid monohydrate
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
38640.01
38640.02
38640.03
SERA38640.01EA
19.56
GBP
SERA38640.01
SERA38640.02
SERA38640.03
Citric acid monohydrate
Citric acid monohydrate
Formula:
C₆H₈O₇.H₂O MW: 210.14 g/mol Melting Pt: 135…152 °C Density: 1.552 g/cm³ (20 °C) Flash Pt: 173.9 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 EINECS: 201-069-1 Merck Index: 13,02350 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...