(S)-(+)-Epichlorhydrin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023907-25G
023907-5G
FLUO023907-25GEA
0
GBP
FLUO023907-25G
FLUO023907-5G
(S)-(+)-Epichlorhydrin
(S)-(+)-Epichlorhydrin
Formula:
ClCH₂CH(O)CH₂ Boiling Pt: 113…115 °C (1013 hPa) Melting Pt: –48 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00077760 CAS Number: 67843-74-7 UN: 2023 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...