(R)-(-)-Epichlorhydrin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
023906-25G
023906-5G
FLUO023906-25GEA
0
GBP
FLUO023906-25G
FLUO023906-5G
(R)-(-)-Epichlorhydrin
(R)-(-)-Epichlorhydrin
Formula:
ClCH₂CH(O)CH₂ Boiling Pt: 115…116 °C (1013 hPa) Melting Pt: –48 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00077759 CAS Number: 51594-55-9 EINECS: 424-280-2 UN: 2023 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...