Hydrochloric acid 2 mol/l (2 N)
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
T134.1
T134.2
ROTHT134.1EA
22.52
GBP
ROTHT134.1
ROTHT134.2
Hydrochloric acid 2 mol/l (2 N)
Hydrochloric acid
Formula:
HCl Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...