(±)-1,2,4-Butanetriol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

19040-100ML 19040-25ML
SIAL19040-100MLEA 182 GBP
SIAL19040-100ML SIAL19040-25ML
(±)-1,2,4-Butanetriol, Sigma-Aldrich®
(±)-1,2,4-Butanetriol
Formula: C₄H₁₀O₃
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.21 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002929
CAS Number: 3068-00-6
EINECS: 221-323-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR