Methanol-[D₄], dehydrated (max. 0.03% H₂O) (99.8% D) for NMR spectroscopy

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

D024H.100ML D024E.10ML
FLUOD024H.100MLEA 0 GBP
FLUOD024H.100ML FLUOD024E.10ML
Methanol-[D₄], dehydrated (max. 0.03% H₂O) (99.8% D) for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
Boiling Pt: 65.3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR