2-Methyl-2-pentenal, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methylpent-2-enal
M-Clarity™ quality level = MQ100
294667-25G
294667-100G
SIAL294667-25GEA
71.8
GBP
SIAL294667-25G
SIAL294667-100G
2-Methyl-2-pentenal, Sigma-Aldrich®
2-Methyl-2-pentenal
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 135…138 °C (1013 hPa) Density: 0.855 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006978 CAS Number: 623-36-9 EINECS: 210-789-5 UN: 1988 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...