Copper, alloy 101, foil, Oxygen-Free High Conductivity (OFHC), Thickness: 1.6 mm (0.064 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

45168.HB 45168.GT
45168.HBEA 231 GBP
45168.HB 45168.GT
Copper, alloy 101, foil, Oxygen-Free High Conductivity (OFHC), Thickness: 1.6 mm (0.064 in)
Copper, alloy 101

Odour: Odourless MDL No.: MFCD00010965

Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR