Phosphorus, red, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
04004-1KG
04004-500G
04004-250G
SIAL04004-1KGEA
131
GBP
SIAL04004-1KG
SIAL04004-500G
SIAL04004-250G
Phosphorus, red, Sigma-Aldrich®
Phosphorus
Formula:
P MW: 30.97 g/mol Density: ∼2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133771 CAS Number: 7723-14-0 EINECS: 231-768-7 UN: 1338 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...