CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

5584.1 5584.2
ROTH5584.1EA 0 GBP
ROTH5584.1 ROTH5584.2
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid)
Formula: C₉H₁₉NO₄S
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00041778
CAS Number: 73463-39-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR