Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% (RT), purum p.a., Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate

M-Clarity™ quality level = MQ200

60063-1KG 60063-250G
SIAL60063-1KGEA 296 GBP
SIAL60063-1KG SIAL60063-250G
Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% (RT), purum p.a., Sigma-Aldrich®
Antimony (III) potassium oxitartrate trihydrate
Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
MW: 667.88 g/mol
Melting Pt: 100
Density: 2.6 g/cm³ (20 °C)
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1 III
Merck Index: 13,00706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR