(-)-Cinchonidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (8α,9R)-Cinchonan-9-ol

M-Clarity™ quality level = MQ200

C80407-100G C80407-10G
SIALC80407-100GEA 162 GBP
SIALC80407-100G SIALC80407-10G
(-)-Cinchonidine, Sigma-Aldrich®
(-)-Cinchonidine
Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 199…202 °C
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00006783
CAS Number: 485-71-2
EINECS: 207-622-3
Merck Index: 13,02307

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR