Cleanroom wipes, ValuSeal® 1500 / LP
Supplier: Berkshire
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ValuSeal®
BERKVSLP-0707B-10EA 0 GBP
BERKVSLP-0707B-10 115-0367 115-0539
Cleanroom wipes, ValuSeal® 1500 / LP
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Sealed edge wipes are ideal for cleaning critical tools and machines, designed for use in manufacturing devices sensitive to micron particle contamination, non-volatile residues and ions. Made from 100% continuous filament double-knitted polyester.
  • Very low in particles and fibres
  • Tear- and abrasion-resistant for tough cleaning
  • Excellent absorbency
  • Lot tracing on each pack
  • Laundered in an ISO 4, Class 10 cleanroom
ValuSeal® LP wipes have fine ultrasonic seals, and ValuSeal® 1500 wipes are laser sealed.
Suitable for use in Class 1, ISO 3, M1.5 cleanrooms.
Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.