U-tube viscometers
Supplier: DIXON SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
338001001 338001002 338001003 338001004 338001005 338001006 338001007 338001008
616-3254EA 300 GBP
616-3254 616-3255 616-3256 616-3257 616-3258 616-3259 616-3260 616-3261
U-tube viscometers
Viscometers Capillary Viscometers
The time for the level of the liquid to pass between these marks is proportional to the kinematic viscosity.
  • Upper bulb and bulb lower down on the other arm
  • Two marks (one above and one below the upper bulb) indicate a known volume
Liquid is drawn into the upper bulb by suction, then flowed down through the capillary into the lower bulb.
Certifications: ASTM445-IP 71 and BS 188