Sealing films, cling film
Supplier: Terinex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
291-0020EA 41.3 GBP
291-0020 291-0021
Sealing films, cling film
Seals General Purpose Sealing Films
Cling film stretches in all directions, made of clear plastic. Can be used for sealing tubes, beakers, flasks and bottles. Clings well to glass, ceramics and stainless steel, but will not cling heavily to itself.
  • Odourless, moisture-proof
  • High tensile strength and puncture resistant
Delivery information: Rolls supplied in a dispenser box.