Gel drying systems, BioDesignGelWrap™
Supplier: BIODESIGN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G101 G103A G103B G105 G113
BDESG101EA 160 GBP
BDESG101 BDESG103A BDESG103B BDESG105 BDESG113
Gel drying systems, BioDesignGelWrap™
Gel Dryers Gel Dryers, Air Drying
An easy and economical way to dry and store all types of polyacrylamide and agarose gels, including slab, gradient, 2D, immunodiffusion, immunoelectrophoretic and enzyme activity gels. The gel is simply sandwiched between two wet pieces of BioDesignGelWrap™ on a BioDesignGelFrame™ and either air-dried (thin gels) or dried using a 60 W incandescent lamp (heavier gels).
  • Gel dries in a tough envelope of BioDesignGelWrap™ - transparent, flat and wrinkle-free
  • Colour of stains for proteins (brilliant blue, silver, etc), glycoproteins, nucleic acids, and coloured enzymatic products remains stable for years
  • Optional storage container maintains the moisture content of the BioDesignGelWrap™ for maximum shelf life