Tubing connectors, T-shaped
Supplier: SIMS Portex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
700/160/005 700/160/010
229-0025EA 12.6 GBP
229-0025 229-0026
Tubing connectors, T-shaped
Fittings and Connectors
PP, translucent.
  • Good mechanical and chemical resistance
  • Very good thermal resistance
  • Autoclavable
For connecting three tubes.