Bottles, wide neck, with standard ground sjoints toppers
Supplier: SMITH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2014B/50 2014/250 2014/500 2014/1000
215-1108EA 0 GBP
215-1108 215-1107 215-1109 215-1106
Bottles, wide neck, with standard ground sjoints toppers
Bottles Glass Bottles
Soda lime glass, clear, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Solid, replaceable glass stoppers
Certifications: DIN 12039