Hybridisation bottles
Supplier: BOEKEL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
502-0100 502-0150 502-0225 502-0300 502-0100R 502-0150R 502-0225R 502-0300R
731-0532EA 205 GBP
731-0532 731-0534 731-0536 731-0538 731-0533 731-0535 731-0537 731-0539
Hybridisation bottles
Bottles Hybridisation Bottles/Tubes
Large acrylic bottles are constructed of beta-blocking acrylic. Designed with screw-on cap and leak proof gasket. Glass bottles are supplied with either blue caps and silicone O-Ring or red caps with PTFE liner.