Trimethyl orthoacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

237876-100ML 237876-500ML
SIAL237876-100MLEA 118 GBP
SIAL237876-100ML SIAL237876-500ML
Trimethyl orthoacetate, Sigma-Aldrich®
Trimethyl orthoacetate
Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00008477
CAS Number: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR