Triethyl orthopropionate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

T60607-100ML T60607-500ML
SIALT60607-100MLEA 92.3 GBP
SIALT60607-100ML SIALT60607-500ML
Triethyl orthopropionate, Sigma-Aldrich®
Triethyl orthopropionate
Formula: C₉H₂₀O₃
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 0.8876 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009226
CAS Number: 115-80-0
EINECS: 204-108-0
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR