3-Fluoro-5-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002160-1G 002160-5G 002160-25G
FLUO002160-1GEA 0 GBP
FLUO002160-1G FLUO002160-5G FLUO002160-25G
3-Fluoro-5-(trifluoromethyl)benzonitrile
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 28 °C
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR