2-Fluoro-3-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF633.5 001999-1G 001999-5G 001999-10G
CHMPCFF633.5EA 0 GBP
CHMPCFF633.5 FLUO001999-1G FLUO001999-5G FLUO001999-10G
2-Fluoro-3-(trifluoromethyl)benzonitrile
2-Fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.323 g/cm³ (20 °C)
MDL Number: MFCD00061152
CAS Number: 146070-35-1
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR