4-Chloro-3-(trifluoromethyl)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007839-5G
007839-1G
FLUO007839-5GEA
0
GBP
FLUO007839-5G
FLUO007839-1G
4-Chloro-3-(trifluoromethyl)benzonitrile
4-Chloro-3-(trifluoromethyl)benzonitrile
Formula:
C₈H₃ClF₃N Boiling Pt: 120…125 °C (20 mmHg) Melting Pt: 66…68 °C |
MDL Number:
MFCD00190117 CAS Number: 1735-54-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...