2-Chloro-4-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004384-1G 004384-25G 004384-5G
FLUO004384-1GEA 0 GBP
FLUO004384-1G FLUO004384-25G FLUO004384-5G
2-Chloro-4-(trifluoromethyl)benzonitrile
2-Chloro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MDL Number: MFCD00084939
CAS Number: 1813-33-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR