2-Chloro-5-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003149-5G 003149-25G
FLUO003149-5GEA 0 GBP
FLUO003149-5G FLUO003149-25G
2-Chloro-5-(trifluoromethyl)benzonitrile
2-Chloro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
Boiling Pt: 210…212 °C (1013 hPa)
Melting Pt: 39…40 °C
Density: 1.48 g/cm³ (20 °C)
MDL Number: MFCD00019742
CAS Number: 328-87-0
EINECS: 206-338-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR