Stainless Steel Pails
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PAI4015JT PAI4015J
COLDPAI4015JTEA 0 GBP
COLDPAI4015JT COLDPAI4015J
Stainless Steel Pails
Buckets
A range of hygienic, high-quality stainless steel pails which meet international pharma and food quality compliance regulations.
  • Pails available as standard, off-the-shelf for delivery within days
  • 304 grade stainless steel
  • Exceptional average surface finish of 0,4 µm Ra
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional material grade, surface finish, and TSE certification available upon request
Made from 304 grade stainless steel, these pails come complete with a base skirtband and a swing handle. They feature crevice-free and pit-free interiors and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information.