ChemFAST ELITE Laboratory Fume Cupboards
Supplier: FASTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FAS/1244/24E15 FAS/1244/24E9
FASTFAS/1244/24E15EA 43210 GBP
FASTFAS/1244/24E15 FASTFAS/1244/24E9
ChemFAST ELITE Laboratory Fume Cupboards
Fume Hoods and Enclosures Ductless Fume Hoods
ChemFAST cabinet may be optionally fitted with water tap and drain system.
  • Power on/off
  • Light on/off
  • Microprocessor monitoring system checking airflow, pre-filter and filter efficiency
  • Variable speed air regulation
  • Audible and visual alarms alert the operator to low/high airflow, fan failure, filter and pre-filter condition, black-out, gas detector and anemometer failure
Work surface and all drain connections are made in AISI 304L stainless steel.
Water tap can be positioned on the right or on the left hand side.
The unit fitted with optionalMore Product Information