Heavy Duty Measuring Jugs
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JUMC0.5G JUM4001G JUM4002G
COLDJUMC0.5GEA 325 GBP
COLDJUMC0.5G COLDJUM4001G COLDJUM4002G
Heavy Duty Measuring Jugs
Beakers
Stainless steel measuring jugs meet international pharma quality compliance regulations.
  • All sizes available as standard, off-the-shelf for delivery within days
  • 304 pharmaceutical grade stainless steel
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Material Grade, Surface Finish, and TSE certification available upon request
Made from 304 grade stainless steel, these measuring jugs are straight sided, graduated, and come with a pouring spout and closed side handle. They feature crevice-free and pit-free interiors and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. Measuring jugs are available in Heavy Duty, Light Duty, and Light Duty with rimmed foot formats.