Stainless Steel Vessels
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VES4020JT VES4030JT VES4050JT VES4020P/V4020 VES4030J VES4050J
COLDVES4020JTEA 0 GBP
COLDVES4020JT COLDVES4030JT COLDVES4050JT COLDVES4020P/V4020 COLDVES4030J COLDVES4050J
Stainless Steel Vessels
Sample Containers General Purpose Containers
Hygienic, durable stainless steel storage, mixing, and transportation containers, available in 304 grade material with a range of modifications to meet every requirement.
  • 304 grade stainless steel
  • Side carrying handles and skirtband as standard
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Available off-the-shelf for immediate dispatch
  • Durable, easy to sterilise, and re-useable, for long service life and greater ROI
  • Exceptional average surface finish of 0,4 micrometers Ra; leaves no pits or crevices for bacteria to store
  • Full material traceability for international pharma-quality compliance
International cleanroom quality, for even the most demanding environments in the pharmaceutical, cosmetics, food and chemical industries.
Lids, airtight toggle clamps and seals, and mobile frames for decanting, are available for all sizes of containers, along with a range of other optional modifications to meet every requirement.