Stainless Steel Containers with Lid
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CON4100PCO4100 CON4150PCOL41 CON4200PCO4200
COLDCON4100PCO4100EA 0 GBP
COLDCON4100PCO4100 COLDCON4150PCOL41 COLDCON4200PCO4200
Stainless Steel Containers with Lid
Sample Containers General Purpose Containers
A range of hygienic, high-quality stainless steel containers which meet international pharma and food quality compliance regulations.
  • Standard sizes available for off-the-shelf for delivery within days
  • Exceptional average surface finish of 0.4 micrometers Ra
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional material grade and surface finish certification available upon request
These containers are available in 304 grade stainless steel, and have a lid with a centre fixed knob. They have crevice-free andMore Product Information