Thymol blue sodium salt 0,1% in ethanol (20%) Reag. Ph. Eur. 1090601
Supplier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium
Ordering information: Normally manufactured to order. Please check with customer services.
Formula:
C₂₇H₂₉NaO₅S |
MDL Number:
MFCD00151093 CAS Number: 62625-21-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...