1-Iodoperfluorodecane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Henicosafluoro-10-iododecane, Perfluorodecyl iodide, Perfluoro-1-iododecane

003284-25G 003284-5G
FLUO003284-25GEA 0 GBP
FLUO003284-25G FLUO003284-5G
1-Iodoperfluorodecane
1-Iodoperfluorodecane
Formula: C₁₀F₂₁I
MDL Number: MFCD00001065
CAS Number: 423-62-1
EINECS: 207-030-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR