3-Fluoro-1-propanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010306-5G
010306-25G
FLUO010306-5GEA
0
GBP
FLUO010306-5G
FLUO010306-25G
3-Fluoro-1-propanol
3-Fluoro-1-propanol
Formula:
C₃H₇FO MW: 78.09 g/mol |
MDL Number:
MFCD00039548 CAS Number: 462-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...