3-Bromo-2-thenoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-2-thiophenecarboxylic acid, 3-Bromothiophene-2-carboxylic acid

019423-25G 019423-5G
FLUO019423-25GEA 0 GBP
FLUO019423-25G FLUO019423-5G
3-Bromo-2-thenoic acid
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR