3,5-Bis(trifluoromethyl)benzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006010-5G
006010-25G
FLUO006010-5GEA
0
GBP
FLUO006010-5G
FLUO006010-25G
3,5-Bis(trifluoromethyl)benzoic acid
3,5-Bis(trifluoromethyl)benzoic acid
Formula:
C₉H₄F₆O₂ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 140…144 °C Density: 1,42 g/cm³ (20 °C) |
MDL Number:
MFCD00000388 CAS Number: 725-89-3 EINECS: 211-970-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...