Uric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H-Purine-2,6,8(3H,7H,9H)-trione
M-Clarity™ quality level = MQ100
U2625-25G
U2625-100G
U2625-1KG
SIALU2625-25GEA
66.9
GBP
SIALU2625-25G
SIALU2625-100G
SIALU2625-1KG
Uric acid, Sigma-Aldrich®
Uric acid
Formula:
C₅H₄N₄O₃ MW: 168.11 g/mol Melting Pt: 436 °C Density: 1.89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005712 CAS Number: 69-93-2 EINECS: 200-720-7 Merck Index: 12,10014 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...