2-(Diisopropylamino)ethanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

471488-100ML 471488-500ML
SIAL471488-100MLEA 47.6 GBP
SIAL471488-100ML SIAL471488-500ML
2-(Diisopropylamino)ethanol, Sigma-Aldrich®
2-(Diisopropylamino)ethanol
Formula: C₈H₁₉NO
Boiling Pt: 187…192 °C (1013 hPa)
Density: 0.826 g/cm³ (25 °C)
CAS Number: 96-80-0
EINECS: 202-536-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR