1-(2-Thenoyl)-3,3,3-trifluoroacetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002361-25G 002361-100G 002361-250G
FLUO002361-25GEA 0 GBP
FLUO002361-25G FLUO002361-100G FLUO002361-250G
1-(2-Thenoyl)-3,3,3-trifluoroacetone
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formula: C₈H₅F₃O₂S
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 40…47 °C
Flash Pt: 111 °C
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR