1-Iodoperfluorohexane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007067-100G
007067-500G
007067-25G
FLUO007067-100GEA
0
GBP
FLUO007067-100G
FLUO007067-500G
FLUO007067-25G
1-Iodoperfluorohexane
1-Iodoperfluorohexane
Formula:
C₆F₁₃I Boiling Pt: 117 °C (1013 hPa) Melting Pt: –45 °C Density: 2,0564 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001063 CAS Number: 355-43-1 EINECS: 206-586-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...