1-Bromo-3,3,3-trifluoro-2-propanol 96%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromo-1,1,1-trifluoropropan-2-ol, 3-Bromo-1,1,1-trifluoro-2-propanol
L10566.03
L10566.06
L10566.03EA
37.7
GBP
L10566.03
L10566.06
1-Bromo-3,3,3-trifluoro-2-propanol 96%
1-Bromo-3,3,3-trifluoro-2-propanol
Purity: 96%
CAS No.: 431-34-5
Molecular Formula: C3H4BrF3O
Formula Weight: 192.97
Flash Point: 43°(109°F)
Refractive Index: 1.3990
Boiling Point: 124-126°
Beilstein Reference: 1737803
UN No.: 1987
MDL No.: MFCD00040945
Formula:
BrH₂CCH(OH)CF₃ MW: 192.96 g/mol Boiling Pt: 124…126 °C Density: 1.861 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040945 CAS Number: 431-34-5 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...