1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,1,1,3,3,3-Hexafluoro-2-methylpropan-2-ol, Hexafluoro-tert-butanol, Hexafluoro-2-methylisopropanol
L09245.06
L09245.14
L09245.06EA
51.7
GBP
L09245.06
L09245.14
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥98%
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Purity: 98%
CAS No.: 1515-14-6
Molecular Formula: C4H4F6O
Formula Weight: 182.07
Refractive Index: 1.2950
Boiling Point: 60-62°
MDL No.: MFCD00039259
Formula:
C₄H₄F₆O MW: 182.07 g/mol Boiling Pt: 60…62 °C Density: 1.484 Storage Temperature: Ambient |
MDL Number:
MFCD00039259 CAS Number: 1515-14-6 EINECS: 216-155-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...