Primesep C HPLC Columns
Supplier: SIELC Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C-32.250.0310 100-21.150.031 MP-21.250.0530
SIELC-32.250.0310EA 1700 GBP
SIELC-32.250.0310 SIEL100-21.150.031 SIELMP-21.250.0530
Primesep C HPLC Columns
Chromatography Columns
The Primesep C mixed-mode HPLC phase from SIELC Technologies has a complexing group which can form a weak complex with amines and metal ions.
  • Improves the retention of positively charged compounds by forming a host-guest complex
  • Retains neutral compounds through reversed-phase mechanisms
  • Mobile phases are LC-MS compatible