White spirit

Supplier: BARRETTINE INDUSTRIAL
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

WHSB025 WHSB205
BARRWHSB025EA 0 GBP
BARRWHSB025 BARRWHSB205
White spirit
White Spirit
CAS Number: 8052-41-3
EINECS: 232-489-3
UN: 1268
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR