Phenyl-β-D-glucopyranoside, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

292710-25G 292710-5G
SIAL292710-25GEA 353 GBP
SIAL292710-25G SIAL292710-5G
Phenyl-β-D-glucopyranoside, Sigma-Aldrich®
Phenyl β-D-glucopyranoside
Formula: C₁₂H₁₆O₆
Boiling Pt: 437 °C (1013 hPa)
Melting Pt: 176 °C
MDL Number: MFCD00064089
CAS Number: 1464-44-4
EINECS: 215-978-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR