Troclosene sodium, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Sodium dichloroisocyanurate, Dichloro-1,3,5-triazinetrione sodium salt, Dichloroisocyanuric acid sodium salt

M-Clarity™ quality level = MQ200

218928-25G 218928-500G
SIAL218928-25GEA 51.6 GBP
SIAL218928-25G SIAL218928-500G
Troclosene sodium, Sigma-Aldrich®
Troclosene sodium
Formula: C₃Cl₂N₃NaO₃
MW: 219.95 g/mol
Melting Pt: 225
Density: 0.96 g/cm³ (20 °C)
MDL Number: MFCD00006036
CAS Number: 2893-78-9
EINECS: 220-767-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR